missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution, 0.1M, Chem-Lab

Product Code. 11913173
Change view
Click to view available options
Solvent or Matrix:
Ethanol
IPA (2-Propanol)
Water
pH:
13
14
Unit Size:
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Solvent or Matrix pH unitSize
11913173 Water 13 1 L
11903193 Ethanol 14 1 L
11983193 IPA (2-Propanol) 13 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11913173 Supplier Chem-Lab Supplier No. CL05.1105.1000
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Used as a titrant in volumetric analysis. Potassium Hydroxide Solution, 0.1M, Chem-Lab is a laboratory reagent that has 5.612g KOH per 1L of solvent. It is highly acidic in nature and is highly corrosive when handled. Choice of alcohol solvents or water matrix.

Potassium hydroxide solution is an effective conductor and alcoholic solutions are highly flammable. It is used in the production of other potassium compounds and as an electrolyte in alkaline batteries, such as those used in television remote controls. It is used in the manufacture of soft soaps and the production of biodiesel. It can be also be used in disinfection agents for surfaces that resist KOH corrosion, in chemical cremation and to identify some species of fungi.

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

pH 13
Color Colorless
Boiling Point 100°C
Melting Point 0°C
Concentration 5.612g KOH/1L solvent
Density 1.01g/mL
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Solubility Information Soluble in Water
Quantity 1 L
IUPAC Name potassium;hydroxide
Formula Weight 56.11g/mol
Normality 0.1 N (±0.0002/20°C)
Odor Odorless
Solvent or Matrix Water
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Potassium hydroxide 0.1 mol/l
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: KOH/H2O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.