missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, 0.500 Normal (N/2) in Ethanol, Ricca
Description
- Pot Hydroxide, 0.5 N in EtOH 120 mL Poly natural
Specifications
Specifications
| CAS | 64-17-5, 7732-18-5, 1310-58-3, 107-15-3 |
| Quantity | 120 mL |
| Grade | ACS |
| Packaging | Natural Poly Container |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?