Learn More
Potassium Hydroxide, 4M, Chem Lab™
Product Code. 11905049
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11905049 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11905049 Supplier Chem-Lab Supplier No. CL05.1132.5000
  • ONLINE EXCLUSIVE PRICE
    121.00€
    57.70€ / 5 L
    Save 63.30€
    52% Off
 Restricted Item
In Stock
Specifications
Color Colorless
Concentration 224g KOH/l H2O = 4 N (±0.008/20°C)
Density 1.17g/mL
Solubility Information Soluble in Water
Quantity 5 L
Odor Odorless
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Potassium hydroxide 4 mol/l
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Potassium hydroxide