missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.01M in IPA, Chem-Lab

Product Code. 11906836
Change view
Click to view available options
Quantity:
1 L
Packaging:
946mL
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11906836 1 L 946mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11906836 Supplier Chem-Lab Supplier No. CL05.1151.1000
  • ONLINE EXCLUSIVE PRICE
    134.00€
    42.40€ / 1 L
    Save 91.60€
    68% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Used as a titrant in volumetric analysis, TMAR. Potassium Hydroxide, 0.01M in IPA, Chem-Lab is a laboratory reagent that has 0.5612g KOH per 1L of 2-Propanol. It is highly acidic in nature and is highly corrosive when handled.

Alcoholic potassium hydroxide is an effective conductor of heat and highly flammable. It is used as a dessicant. It is also used in some electrical batteries, such as those used in television remote controls. It is used in the manufacture of soaps, the production of biodiesel and the production of other potassium compounds.

Specifications

pH 13 (20°C, H2O)
Color Colorless
CAS 1310-58-3, 67-63-0
Molecular Formula HKO
Concentration 0.5612g KOH per 1L C3H8O (0.01mol/L)
MDL Number MFCD00003553
Density 0.78g/mL
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Solubility Information Soluble in water
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[K+]
IUPAC Name potassium;hydroxide
Explosion Limit Lower 2% Volume/Upper 12% Volume
Molecular Weight (g/mol) 56.11
Flash Point 12°C (IPA)
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 60.10g/mol
Normality 0.01N (±0.0002, 20°C)
Odor 2-propanol
Packaging 946mL
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium hydroxide 0.01 mol/l in Propanol-2
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydroxide,propan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.