Learn More
Potassium Hydroxide Solution, 0.1M, Chem Lab™
Product Code. 11933173
Change view
Click to view available options
Quantity:
10 L
Packaging:
10L
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11933173 10 L 10L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11933173 Supplier Chem-Lab Supplier No. CL05.1105.9010
  • ONLINE EXCLUSIVE PRICE
    273.00€
    106.00€ / 10 L
    Save 167.00€
    61% Off
 Restricted Item
In Stock
Specifications
Color Colorless
Concentration 5.611g KOH/l H2O = 0.1 N (±0.0002/20°C)
Density 1.01g/mL
Solubility Information Soluble in water
Quantity 10 L
Odor Odourless
Packaging 10L
Physical Form Liquid
compliance-icons
Product Identifier
  • Potassium hydroxide 0.1 mol/l
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: KOH/H2O