Learn More
Potassium Hydroxide Solution, 0.5M, Chem Lab™
Product Code. 11993183
Change view
Click to view available options
Quantity:
1 L
Packaging:
1L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11993183 1 L 1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11993183 Supplier Chem-Lab Supplier No. CL05.1124.1000
  • ONLINE EXCLUSIVE PRICE
    129.00€
    41.90€ / 1 L
    Save 87.10€
    67% Off
 Restricted Item
In Stock
Specifications
Color Colorless
Concentration 28.055g KOH/l C2H5OH = 0.5 N (±0.001/20°C)
Density 0.85g/mL
Solubility Information Soluble in water
Quantity 1 L
Explosion Limit Lower 2% Volume/Upper 12% Volume
Odor Alcoholic
Packaging 1L
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Potassium hydroxide 0.5 mol/l in Ethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, potassium hydroxide