missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate solution 0.016mol/L, Fisher Chemical™

Product Code. 12983584 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
This item is not returnable. View return policy

Product Code. 12983584

Brand: Fisher Chemical VL 271 1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
CAS 7758-05-6, 7732-18-5
Molecular Formula IKO3
Concentration 0.0167 to 0.0167 M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES [K+].[O-][I](=O)=O
IUPAC Name potassium iodate
Molecular Weight (g/mol) 214.00
PubChem CID 23665710
Quantity 1000 mL
Chemical Name or Material Potassium Iodate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.