missing translation for 'onlineSavingsMsg'
Learn More

Potassium Osmate(VI) Dihydrate, TRC

Product Code. 30411534
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411534 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411534 Supplier Toronto Research Chemicals Supplier No. TRCP6981601G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Osmate(VI) Dihydrate
CAS 10022-66-9
Quantity 1 g
Molecular Formula H4K2O6Os . 2H2O
Synonym (T-4)-Dipotassium Osmate (OsO42-) Dihydrate, Osmic Acid (H2OsO4) Dipotassium Salt Dihydrate, Dipotassium Tetrahydroxodioxoosmate, Dipotassium Tetrahydroxodioxoosmate(2-), Dipotassium Tetrahydroxydioxoosmate(2-), Dipotassium trans-Tetrahydroxodioxoosmate(2-), Potassium Osmate (K2OsO2(OH)4), Potassium Osmate (K2OsO4) Dihydrate, Potassium Osmate Dihydrate, Potassium Osmium Oxide Hydroxide (K2OsO2(OH)4), Potassium Tetrahydroxodioxoosmate (K2[OsO2(OH)4]), trans-Dipotassium Tetrahydroxodioxoosmate(2-), trans-Potassium Tetrahydroxodioxoosmate(2-)
InChI Formula InChI=1S/2K.2H2O.4O.Os/h;;2*1H2;;;;;/q2*+1;;;;;2*-1;
SMILES O.O.[K+].[K+].[O-][Os](=O)(=O)[O-]
IUPAC Name dipotassium;dioxido(dioxo)osmium;dihydrate
Molecular Weight (g/mol) 368.46
Formula Weight 369.8897
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.