missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Dibasic, MP Biomedicals™

Product Code. 11417202 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417202 100 g 100g
11427202 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417202 Supplier MP Biomedicals Supplier No. 02151945.1
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Used as a component for a wide variety of media used in the culture of microorganisms and in phosphate buffered saline (PBS). Potassium Phosphate Dibasic, MP Biomedicals is a reagent with high buffering capacity. In addition to helping maintain pH, it sup

Potassium phosphate occurs in several forms: monobasic, dibasic and tribasic. Most pH neutral potassium phosphate buffer solutions consist of mixtures of the monobasic and dibasic forms to varying degrees, depending on the desired pH.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Soluble in water (1g/20mL); Insoluble to slightly soluble in ethanol; insoluble in alcohol.
Formula Weight 174.18
Percent Purity ≥98% (Anhydrous)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.