missing translation for 'onlineSavingsMsg'
Learn More

Melford Potassium Phosphate, Monobasic

Product Code. 30018710
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018710 2.5 kg 2.5kg
30018709 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018710 Supplier Melford Supplier No. P412002500.0
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potassium Phosphate, Monobasic, ACS Grade is a chemical compound that holds significance in the life science research, particularly in molecular biology, cell culture, and biochemistry fields.

It is a high-purity reagent with ACS (American Chemical Society) grade certification

Potassium Phosphate, Monobasic, ACS Grade is primarily utilized as a component in laboratory solutions and buffers. It is commonly used as a source of potassium ions and as a pH regulator in various experimental procedures. It is particularly effective in applications requiring slightly acidic to neutral pH conditions. Also, It can be used in protein purification, enzyme assays, nucleic acid research, and other biochemical and molecular biology techniques.

Specifications

Chemical Name or Material Potassium Phosphate, Monobasic, ACS Grade
CAS 7778-77-0
Quantity 2.5 kg
Molecular Formula KH2PO4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.