missing translation for 'onlineSavingsMsg'
Learn More

Potassium Standard Solution (Plasma HIQU), Chem Lab™

Product Code. 11919082
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11919082 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11919082 Supplier Chem-Lab Supplier No. CL01.1101.0100
  • ONLINE EXCLUSIVE PRICE
    76.00€
    37.20€ / 100mL
    Save 38.80€
    51% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Concentration 1000 μg/mL
Quantity 100 mL
compliance-icons
Product Identifier
  • Potassium standard solution 10 µg/ml
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Nitric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.