missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Superoxide, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Potassium Superoxide |
| CAS | 12030-88-5 |
| Molecular Formula | KO2 |
| Quantity | 10 g |
| Synonym | Potassium Dioxide, Potassium Oxide |
| InChI Formula | InChI=1S/K.O2/c;1-2/q+1;-1 |
| SMILES | [K]O[O] |
| IUPAC Name | λ^{1}-oxidanyloxypotassium |
| Molecular Weight (g/mol) | 71.1 |
| Formula Weight | 70.9535 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction