missing translation for 'onlineSavingsMsg'
Learn More

Potassium tert-pentyloxide, 14-18% w/v in cyclohexane

Product Code. 11304108
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304108 5 g 5g
11324108 25 g 25g
11314108 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11304108 Supplier Thermo Scientific Alfa Aesar Supplier No. 041394.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Coreactant for conversion of ketones to oxides | CAS: 41233-93-6 | C5H11KO | 126.24 g/mol

Potassium tert-pentyloxide is used in wide range of applications in organic synthesis, agrochemicals, pharmaceuticals, colorants and aroma chemicals. It is also used in manufacturing detergents and biodiesel. They also act as catalysts in polymerization and isomerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41233-93-6
Molecular Formula C5H11KO
Molecular Weight (g/mol) 126.24
MDL Number MFCD00064808
InChI Key ZRLVQFQTCMUIRM-UHFFFAOYSA-N
Synonym potassium 2-methylbutan-2-olate, potassium tert-amylate, potassium tert-pentoxide, potassium tert-pentylate, potassium tert-pentoxide solution, potassium 2-methyl-2-butoxide, 2-butanol, 2-methyl-, potassium salt, potassium tert-amyloxide, 2-butanol, 2-methyl-, potassium salt 1:1
PubChem CID 23683543
SMILES [K+].CCC(C)(C)[O-]

Specifications

Density 0.80g/mL
Flash Point −18° (0°F)
Linear Formula CH3CH2C(CH3)2OK
Quantity 5 g
UN Number UN2924
Sensitivity Air & Moisture Sensitive
IUPAC Name potassium;2-methylbutan-2-olate
Formula Weight 126.24
Concentration or Composition (by Analyte or Components) 14 to 18% w/v in cyclohexane
Physical Form Liquid
Chemical Name or Material Potassium tert-pentyloxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.