missing translation for 'onlineSavingsMsg'
Learn More

Potassium trifluoro(3-nitrophenyl)borate, 95%, BLD Pharm™

Product Code. 30798343
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30798343 1 g 1g
30798032 10 g 10g
30786823 5 g 5g
30787916 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30798343 Supplier BLD Pharm Supplier No. BD1486701g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 192863-40-4
Molecular Formula C6H4BF3KNO2
MDL Number MFCD03701627
Quantity 1 g
SMILES F[B-](F)(C1=CC=CC([N+]([O-])=O)=C1)F.[K+]
Molecular Weight (g/mol) 229.01
Percent Purity 95%
Chemical Name or Material Potassium trifluoro(3-nitrophenyl)borate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.