missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Trifluoroacetate, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | Potassium Trifluoroacetate |
| CAS | 2923-16-2 |
| Molecular Formula | C2F3KO2 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C2HF3O2.K/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1 |
| SMILES | [K+].[O-]C(=O)C(F)(F)F |
| IUPAC Name | potassium;2,2,2-trifluoroacetate |
| Molecular Weight (g/mol) | 152.11 |
| Formula Weight | 151.9487 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction