Learn More
POTEG Monoclonal Antibody (OTI10A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808186S
Description
POTEG Monoclonal antibody specifically detects POTEG in Human samples. It is validated for Western BlotSpecifications
| POTEG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P01375 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7124 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI10A6 | |
| Unconjugated | |
| Tnf | |
| A26C2; ACTBL1; ANKRD26-like family C member 2; ANKRD26-like family C, member 2; cancer/testis antigen family 104, member 4; CT104.4; POTE ankyrin domain family member G; POTE ankyrin domain family, member G; POTE14; POTE-14; POTE14alpha; POTE22; POTEG; Prostate, ovary, testis-expressed protein on chromosome 14; protein expressed in prostate, ovary, testis, and placenta 14 | |
| Full length human recombinant protein of human TNF produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.