Learn More
POTEG Monoclonal Antibody (OTI10A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808186
Encompass_Preferred
 View more versions of this product
Product Code. 17599153
  • 540.87€ / 100µL
Description
Belongs to the POTE family. 3 isoforms of the human protein are produced by alternative splicing. Bera T.K., Gene 337:45-53(2004).
Specifications
Specifications
POTEG
Monoclonal
Unconjugated
POTEG
A26C2, ACTBL1, CT104.4, POTE-14, POTE14, POTE14alpha, POTE22
Mouse
Affinity Chromatography
RUO
404785
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI10A6
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q6S5H5
POTEG
Full length human recombit protein of human POTEG produced in HEK293T cell.