Learn More
Ohaus™ Power Adapter
Product Code. 11782148 Shop All Ohaus Products
Change view
Click to view available options
:
Voltage: 120V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11782148 Voltage: 120V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11782148 Supplier Ohaus™ Supplier No. 46001724
Use this universal power adaptor with Ohaus™ balances.
  • Without plug
  • 120V, 50/60Hz
TRUSTED_SUSTAINABILITY
Specifications
Electrical Requirements 120 V 50/60 Hz
Quantity 1 Each
Type Power Adapter