Learn More
3M™ Powered and Supplied Air Breathing Tube, S-222
Product Code. 31297050 Shop All 3M Products
Change view
Click to view available options
Material:
Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material
31297050 Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31297050 Supplier 3M™ Supplier No. 7100358220
  • ONLINE EXCLUSIVE PRICE
    452.00€
    295.42€ / Pack of 1
    Save 156.58€
    34% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Replacement Part part of a CE-approved respiratory system.
  • Connects the air source with the headtop for reliable, clean, and unrestricted airflow.
  • Compatible with selected 3M™ Respiratory Headtops and Air Delivery Units.
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Disposable Single-use
Material Plastic
Product Type Breathing Tube Installation Accessory
Size One Size