Learn More
Fisherbrand™ PowerPro 3 Amp
Product Code. 15828481 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Display:
2.4 in. TFT-LCD
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Unit Size
15828481 2.4 in. TFT-LCD Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15828481
  • ONLINE EXCLUSIVE PRICE
    1365.00€
    862.36€ / Pack of 1
    Save 502.64€
    36% Off
Estimated Shipment: 05-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Fisherbrand™ PowerPro 3 Amp is a power supply designed for virtually all high current electrophoresis applications.
  • 5 pairs of outputs
  • Higher current output capability is perfect for electroblotting units with high-intensity plate electrodes, particularly Cleaver Scientific omniBLOT maxi, VS20 WAVE and semi dry blotting systems.
  • Electrotransfers may be performed as timed runs in constant or programmable mode to prevent overheating and buffer depletion.
  • A run time extendable to a maximum 999 minutes in constant mode is also useful for overnight transfers undertaken at constant low current in wire electrode systems.
  • Shares the built-in protocol and constant parameter features of the other PowerPro models.
Specifications
For Use With (Equipment) Electroblotting units with high-intensity plate electrodes
Timer Constant mode: 9999 (min.) with alarm Programmable mode: 999 (min.) with alarm
Display 2.4 in. TFT-LCD
Voltage 100 to 240 V rated, 5 to 300 V output, 1 V increments
Wattage 300 w output, 1 w increments
Amperage 10 to 3000 mA output, 10 mA increments