missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PP11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569162
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 92%; Dog: 75%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 83%; Rat: 100%.
This gene encodes a protein with protease activity and is expressed in the placenta. The protein may be useful as a tumor marker. Multiple alternatively spliced transcript variants have been found for this protein.
Specifications
| PP11 | |
| Polyclonal | |
| Unconjugated | |
| ENDOU | |
| 22 serine protease; 26 serine protease; endonuclease, poly(U) specific; endonuclease, poly(U)-specific; endonuclease, polyU-specific; ENDOU; P11; placental protein 11; placental protein 11 related; placental protein 11-related protein; Poly(U)-specific endoribonuclease; PP11; Pp11r; PP11-related protein; Protein endoU; PRSS26; T-cell-specific protein 30; Tcl-30; Uridylate-specific endoribonuclease | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19011 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q3V188 | |
| ENDOU | |
| synthetic peptide directed towards the middle region of human PP11 (aa 179-228). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction