missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18002-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein with protease activity and is expressed in the placenta. The protein may be useful as a tumor marker. Multiple alternatively spliced transcript variants have been found for this protein.Specifications
| PP11 | |
| Polyclonal | |
| Unconjugated | |
| ENDOU | |
| 26 serine protease, ENDOU, P11, Placental protein 11, PP11, Protein endoU, PRSS26 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19011, 680317, 8909 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P21128, Q3V188 | |
| Endou | |
| PP11 Fusion Protein Ag12494 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction