missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PP5 Monoclonal Antibody (24GB6210)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA557616
This item is not returnable.
View return policy
Description
PP5 Monoclonal Antibody for Western Blot
This gene encodes a serine/threonine phosphatase which is a member of the protein phosphatase catalytic subunit family. Proteins in this family participate in pathways regulated by reversible phosphorylation at serine and threonine residues; many of these pathways are involved in the regulation of cell growth and differentiation. The product of this gene has been shown to participate in signaling pathways in response to hormones or cellular stress, and elevated levels of this protein may be associated with breast cancer development. Alternative splicing results in multiple transcript variants.
Specifications
| PP5 | |
| Monoclonal | |
| Unconjugated | |
| Ppp5c | |
| ANP receptor; AU020526; EGK_10788; FLJ36922; FLJ55954; OTTHUMP00000165834; PP5; PPP5; Ppp5c; PPT; PP-T; protein phosphatase 5 catalytic subunit; protein phosphatase 5, catalytic subunit; protein phosphatase T; Serine/threonine-protein phosphatase 5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5536 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 24GB6210 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P53041 | |
| Ppp5c | |
| Recombinant protein of human PPP5C (aa 1-499). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction