Learn More
PPAR alpha/gamma Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA3820
Description
PA3-820 detects peroxisome proliferator activated receptor (PPAR) gamma 2 from human, mouse, and rat tissues and cells. PA3-820 has been successfully used in Western blot, immunoprecipitation and gel shift procedures. By Western blot, this antibody detects an ∽55 kDa protein representing PPAR gamma2 from 3T3-L1 cell lysate. PA3-820 immunizing peptide corresponds to amino acid residues 484-498 from rat PPAR gamma2. This sequence is completely conserved in bovine, chicken, human, mouse, pig and rat PPAR gamma1, PPAR alpha and NUC1.
Peroxisome proliferators are non-genotoxic carcinogens which are purported to exert their effect on cells through their interaction with members of the nuclear hormone receptor family termed peroxisome proliferator activated receptors (PPAR’s). Nuclear hormone receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate ligand. Studies indicate that PPAR’s are activated by peroxisome proliferators such as clofibric acid, nafenopin, and WY-14,643, as well as by some fatty acids. It has also been shown that PPAR’s can induce transcription of acyl coenzyme A oxidase & cytochrome P450 (CYP450) A6 through interaction with specific response elements. PPAR, like several other nuclear hormone receptors, heterodimerizes with retinoid X receptor (RXR) alpha.
Specifications
| PPAR alpha/gamma | |
| Polyclonal | |
| PPARA | |
| Peroxisome Proliferator Activated Receptor | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Gel Shift, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| P37231, Q07869, P23204, P37238, O88275, P37230 | |
| PPARA, PPARG | |
| Synthetic Peptide: I(484) K K T E T D M S L H P L L Q(498) | |
| RUO | |
| 19013, 19016, 25664, 25747, 5465, 5468 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.