missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIL6 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00285755-B01P
This item is not returnable.
View return policy
Description
PPIL6 Polyclonal antibody specifically detects PPIL6 in Human samples. It is validated for Western Blot
Specifications
| PPIL6 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| bA425D10.6, Cyclophilin-like protein PPIL6, dJ919F19.1, EC 5.2.1.8, MGC27054, MGC27056, MGC41939, peptidyl-prolyl cis-trans isomerase-like 6, peptidyl-prolyl cis-trans isomerase-like variant a, peptidylprolyl isomerase (cyclophilin)-like 6, PPIase, radial spoke 12 homolog, Rotamase PPIL6, RSPH12 | |
| PPIL6 (NP_775943.1, 1 a.a. - 311 a.a.) full-length human protein. MARPQPCGPPHARCGSPSLPERPLQVKVVGLFSCPNFQIAKSAAENLKNNHPSKFEDPILVPLQEFAWHQYLQEKKRELKNETWEYSSSVISFVNGQFLGDALDLQKWAHEVWDIVDIKPSALYDALTEDFSAKFLRDTKHDFVFLDICIDSSPIGRLIFELYCDVCPKTCKNFQVLCTGKAGFSQRGIRLHYKNSIFHRIVQNGWIQGGDIVYGKGDNGESIYGPTFEDENFSVPHNKRGVLGMANKGRHSNGSQFYITLQATPYLDRKFVAFGQLIEGTEVLKQLELVPTQNERPIHMCRITDSGDPYA | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| NP_775943.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 285755 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction