missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPM1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12961-1-AP-20UL
This item is not returnable.
View return policy
Description
PPM1A (Protein phosphatase 1A) is also named as PPPM1A and belongs to the PP2C family. It is a serine/threonine protein phosphatase that is essential for regulating cellular stress responses in eukaryotes. PPM1A plays an important role in tumor suppression and that it negatively controls many signaling pathways which are critical for trophoblast invasion. This protein distributes in both the cytoplasm and nucleus of mammalian cells. It has 2 isoforms produced by alternative splicing.Specifications
| PPM1A | |
| Polyclonal | |
| Unconjugated | |
| PPM1A | |
| PP2C ALPHA, PP2CA, PPM1A, PPPM1A, Protein phosphatase 1A, Protein phosphatase IA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19042, 24666, 5494 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P20650, P35813, P49443 | |
| Ppm1a | |
| PPM1A Fusion Protein Ag3645 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction