Learn More
PPM1D Monoclonal Antibody (OTI4H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811163S
Description
PPM1D Monoclonal antibody specifically detects PPM1D in Human, Mouse samples. It is validated for Western BlotSpecifications
| PPM1D | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95831, Q9Z0X1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 26926, 9131 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4H3 | |
| Unconjugated | |
| AIFM1 | |
| AV338790; p53-induced protein phosphatase 1; PP2C delta; PP2CD; PP2C-delta; PPM1D; Protein phosphatase 1D; protein phosphatase 1D magnesium-dependent, delta isoform; protein phosphatase 2C delta isoform; protein phosphatase 2C isoform delta; Protein phosphatase magnesium-dependent 1 delta; protein phosphatase Mg2+/Mn2+ dependent 1D; protein phosphatase Wip1; protein phosphatase, Mg2+/Mn2+ dependent 1D; protein phosphatase, Mg2+/Mn2+ dependent, 1D; wild-type p53-induced phosphatase 1; WIP 1; WIP1 | |
| Human recombinant protein fragment corresponding to amino acids 134-483 of human AIFM1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.