missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R17 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | PPP1R17 |
|---|---|
| Concentration | 850 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktkode | Brand | Quantity | Pris | Mængde & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Mængde & tilgængelighed | |||||
|
30236340
|
Proteintech
27976-1-AP-20UL |
20 μL |
145.00€ 129.00€ / 20µL Save 16.00€ 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235585
|
Proteintech
27976-1-AP-150UL |
150 μL |
410.00€ 349.00€ / 150µL Save 61.00€ 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
The PPP1R17 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PPP1R17. This antibody recognizes human, mouse, rat antigen. The PPP1R17 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Tekniske data
| PPP1R17 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| GSBS | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 850 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 10842 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel