missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R18 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18432B
This item is not returnable.
View return policy
Description
PPP1R18 Polyclonal antibody specifically detects PPP1R18 in Human samples. It is validated for Western Blot
Specifications
| PPP1R18 | |
| Polyclonal | |
| Western Blot | |
| HKMT1098, KIAA1949, phostensin, PPP1R18, Protein Phosphatase 1 F-Actin Cytoskeleton Targeting Subunit, Protein Phosphatase 1 F-Actin Cytoskeleton-Targeting Subunit, Protein Phosphatase 1 Regulatory Subunit 18, Protein Phosphatase 1, Regulatory Subunit 18 | |
| Recognizes region between residues Between Between 325 and 375.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 170954 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction