missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R3C Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-26081PCP
This item is not returnable.
View return policy
Description
PPP1R3C Polyclonal antibody specifically detects PPP1R3C in Human samples. It is validated for Western Blot
Specifications
| PPP1R3C | |
| Polyclonal | |
| Western Blot | |
| Phosphatase 1, regulatory inhibitor subunit 5, PPP1R5PP1 subunit R5, protein phosphatase 1 regulatory subunit 3C, Protein phosphatase 1 regulatory subunit 5, protein phosphatase 1, regulatory (inhibitor) subunit 3C, Protein targeting to glycogen, PTG | |
| Produced in rabbits immunized with E. coli-derived Human PPP1R3C fragment | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 5507 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction