missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PPP2R5A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00€ - 508.00€
Specifications
| Antigen | PPP2R5A |
|---|---|
| Dilution | Western Blot : 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067165
|
Novus Biologicals™
NBP3-30775-100ug |
100 μg |
508.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066964
|
Novus Biologicals™
NBP3-30775-25ug |
25 μg |
213.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPP2R5A Polyclonal antibody specifically detects PPP2R5A in Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
| PPP2R5A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Wnt Signaling Pathway | |
| Phosphate buffered saline | |
| 5525 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1:500-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| B56A, MGC131915, PP2A B subunit isoform B56-alpha, PP2A B subunit isoform B'-alpha, PP2A B subunit isoform PR61-alpha, PP2A B subunit isoform R5-alpha, PP2A, B subunit, B' alpha isoform, PP2A, B subunit, B56 alpha isoform, PP2A, B subunit, PR61 alpha isoform, PP2A, B subunit, R5 alpha isoform, PR61A, PR61alpha, protein phosphatase 2, regulatory subunit B (B56), alpha isoform, protein phosphatase 2, regulatory subunit B', alpha, protein phosphatase 2, regulatory subunit B', alpha isoform, serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, alphaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit alpha isoform | |
| The immunogen used to generate the purified antibody was a peptide conjugated to KLH corresponding to the sequence NH2-Cys-Gly-Glu-Glu-Asp-Ile-Asp-THr-ArgLys-Ile-Asn-Asn-Ser-Phe-COOH. This peptide antibody corresponds to N terminal peptide of PPP2R5A regulatory subunit having a MW of 53kDa. The sequence used is amino acid 2-14. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title