missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PPP2R5C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30779-25ug
This item is not returnable.
View return policy
Description
PPP2R5C Polyclonal antibody specifically detects PPP2R5C in Bovine,Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| PPP2R5C | |
| Polyclonal | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| B' alpha regulatory subunit, B56G, KIAA0044, MGC23064, PP2A B subunit isoform B56-gamma, PP2A B subunit isoform B'-gamma, PP2A B subunit isoform PR61-gamma, PP2A B subunit isoform R5-gamma, PR61G, protein phosphatase 2, regulatory subunit B (B56), gamma isoform, protein phosphatase 2, regulatory subunit B', gamma, protein phosphatase 2, regulatory subunit B', gamma isoform, Renal carcinoma antigen NY-REN-29, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit gamma isoform | |
| Antibody raised against synthetic peptide corresponding to amino acids 53 to 66 of the mammalian protein PPP2R5C conjugated to KLH. | |
| 25 μg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Phosphatase, Signal Transduction, Wnt Signaling Pathway | |
| 5527 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction