missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HPV Type 18 E7 Recombinant Rabbit Monoclonal Antibody (HL2787)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556752
This item is not returnable.
View return policy
Description
HPV Type 18 E7 Recombinant Monoclonal Antibody for Western Blot
Infection with specific types of HPV has been associated with an increased risk of developing cervical neoplasia. HPV types 6 and 11 have been associated with relatively benign diseases such as genital warts but types 16 and 18 are strongly associated with cervical, vaginal, and vulvar malignancies. E7-driven degradation of pRb may be involved in cervical tumorigenesis in humans.
Specifications
| HPV Type 18 E7 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot | |
| HL2787 | |
| Unconjugated | |
| HPV E7; HPV18; HPV-18; human papillomavirus | |
| Full length Human Papillomavirus type 18 E7 recombinant protein. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto