missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP3CA Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55147-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to PPP3CA.
Calcineurin plays an important role in the T-cell receptor-mediated signal transduction pathway. The protein encoded by this gene binds specifically to the activated form of calcineurin and inhibits calcineurin-mediated signal transduction. The encoded protein is found in the nucleus and contains a leucine zipper domain as well as several PEST motifs, sequences which confer targeted degradation to those proteins which contain them. Alternative splicing results in multiple transcript variants encoding two different isoforms.Specifications
| PPP3CA | |
| Polyclonal | |
| Unconjugated | |
| PPP3CA | |
| Calcineurin A alpha, CALN, CALNA, CALNA1, CAM PRP catalytic subunit, CCN1, CNA, CNA1, PPP2B, PPP3CA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19055, 5530 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P63328, Q08209 | |
| Ppp3ca | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction