missing translation for 'onlineSavingsMsg'
Learn More

Prasterone, Mikromol™

Product Code. 18030010
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18030010 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18030010 Supplier Mikromol Supplier No. MM0651.000250
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Prasterone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dehydroisoandrosterone (DHEA)
CAS 53-43-0
Analyte or Component Names Dehydroisoandrosterone (DHEA)
Shipping Condition Room Temperature
Molecular Formula C19 H28 O2
Certifications/Compliance ISO 17034
Synonym 17-Chetovis, 17-Hormoforin, 5,6-Dehydroisoandrosterone, 5,6-Didehydroisoandrosterone, 5-Dehydroepiandrosterone, Androstenolone, Astenile, DHA, DHEA, Deandros, Dehydro-epi-androsterone, Dehydroepiandrosterone, Dehydroisoandrosterone, Diandron, Diandrone, EM 760, GL 701, IM 28, Immunor, Intrarosa, NSC 9896, Prasterone, Psicosterone, Siscelar plus, trans-Dehydroandrosterone, Androst-5-en-17-one, 3-hydroxy-, (3beta)-, Androst-5-en-17-one, 3beta-hydroxy- (8CI), (3beta)-3-Hydroxyandrost-5-en-17-one, 3beta-Hydroxy-Delta5-androsten-17-one, 3beta-Hydroxyandrost-5-en-17-one, 3beta-Hydroxyandrost-5-ene-17-one, 5-Androsten-3beta-ol-17-one, Androst-5-ene-17-one-3beta-ol, Androst-5-ene-3beta-ol-17-one, Delta5-Androsten-3beta-ol-17-one
InChI Formula InChI=1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h3,13-16,20H,4-11H2,1-2H3/t13-,14-,15-,16-,18-,19-/m0/s1
SMILES C[C@]12CC[C@H]3[C@@H](CC=C4C[C@@H](O)CC[C@]34C)[C@@H]1CCC2=O
IUPAC Name (3S,8R,9S,10R,13S,14S)-3-hydroxy-10,13-dimethyl-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-one
Molecular Weight (g/mol) 288.424
Quantity 250 mg
Formula Weight 288.209 g/mol
Show More Show Less
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.