missing translation for 'onlineSavingsMsg'
Learn More

Praziquantel, TRC

Product Code. 30137632
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137632 5 g 5g
17993245 50 mg 50mg
17963045 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137632 Supplier Toronto Research Chemicals Supplier No. TRCP7020955G
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55268-74-1
Molecular Formula C19 H24 N2 O2
Molecular Weight (g/mol) 312.41
Synonym Praziquantel, 4H-Pyrazino[2,1-a]isoquinolin-4-one, 2-(cyclohexylcarbonyl)-1,2,3,6,7,11b-hexahydro-, 2-(Cyclohexylcarbonyl)-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, (±)-Praziquantel, (±)-Praziquatel, 2-(Cyclohexanecarbonyl)-2,3,6,7-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4(11bH)-one, 2-(Cyclohexanecarbonyl)-3,6,7,11b-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4-one, 2-Cyclohexanecarbonyl-1,2,3,6,7,11b-hexahydro-pyrazino[2,1-a]isoquinolin-4-one, Azinox, Benesal, Biltricide, Cesol, Cysticide, Distocide, Droncit, Embay 8440, Epiquantel, Hada Clean, Prazinon, Praziquantel, Pyquiton, Saniquantrel, Warmnil
IUPAC Name (11bR)-2-(cyclohexanecarbonyl)-3,6,7,11b-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4-one
SMILES O=C(C1CCCCC1)N2C[C@@H]3N(CCc4ccccc34)C(=O)C2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C19H24N2O2/c22-18-13-20(19(23)15-7-2-1-3-8-15)12-17-16-9-5-4-6-14(16)10-11-21(17)18/h4-6,9,15,17H,1-3,7-8,10-13H2/t17-/m0/s1
Formula Weight 312.1838
Percent Purity >95%
Chemical Name or Material Praziquantel
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.