missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDM9 Antibody (RAB-C370), DyLight 594 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20141DL594
This item is not returnable.
View return policy
Description
PRDM9 Monoclonal antibody specifically detects PRDM9 in Human samples. It is validated for ELISA, Immunofluorescence
Specifications
| PRDM9 | |
| Monoclonal | |
| DyLight 594 | |
| EC 2.1.1.43, histone-lysine N-methyltransferase PRDM9, minisatellite binding protein 3 (115kD), minisatellite binding protein 3, 115kDa, PFM6MEISETZ, PR domain containing 9, PR domain zinc finger protein 9, PR domain-containing protein 9, PRMD9, ZNF899 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRDM9 protein under non-denaturing conditions. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunofluorescence | |
| RAB-C370 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 56979 | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction