missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20568-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to PRDX1.
This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. Three transcript variants encoding the same protein have been identified for this gene.Specifications
| PRDX1 | |
| Polyclonal | |
| Unconjugated | |
| PRDX1 | |
| MSP23, NKEF A, NKEFA, PAG, PAGA, PAGB, peroxiredoxin 1, PRDX1, PRX1, PRXI, TDPX2, Thioredoxin peroxidase 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117254, 18477, 5052 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.48 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35700, Q06830, Q63716 | |
| Prdx1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction