Learn More
Hanna Instruments™ pH/EC/TDS Multiparameter Probe
Description
- Pre-amplified pH electrode
- Amperometric EC/TDS sensor
- Built-in temperature sensor
Order Info
Includes
Specifications
Specifications
| Range (pH) | 0 to 13 |
| Temperature Range (Metric) | 0°C to 50°C |
| For Use With (Application) | Measures pH, EC/TDS, and temperature in greenhouses, hydroponics, environmental monitoring, water treatment, and boilers and cooling towers |
| Includes | Electrolyte: gel |
| Model | HI-1285-5 |
| Product Type | Preamplified pH and EC Probe |
| Length (Metric) | 185 mm |