Learn More
Epredia™ Precision Cut Paraffin
Product Code. 10698694 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Paraffin
Quantity:
8 x 1 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity Unit Size
10698694 Paraffin 8 x 1 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10698694 Supplier Epredia™ Supplier No. 501006
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Achieve thorough tissue infiltration and provide maximum support during microtomy with high-quality Precision Cut Paraffin. Also available in prepackaged kit for Epredia™ Excelsior™ AS and Revos™ Tissue Processors.
  • Quality blend of paraffin, polymers and plastics (to varying degrees, based upon type)
TRUSTED_SUSTAINABILITY
Specifications
Melting Point 54° to 57°C
Product Type Paraffin
Quantity 8 x 1 kg
For Use With (Application) General purpose