missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prednisone, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Prednisone
Specifications
Specifications
| Chemical Name or Material | Prednisone |
| CAS | 53-03-2 |
| Analyte or Component Names | Prednisone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H26 O5 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Prednisone, 17,21-Dihydroxypregna-1,4-diene-3,11,20-trione, Prednisolone Imp. B (EP), 1,2-Dehydrocortisone, 1-Dehydrocortisone, 17,21-Dihydroxypregn-1,4-diene-3,11,20-trione, Adasone, Ancortone, Apo-Prednisone, Bicortone, Cartancyl, Colisone, Cordrol, Cortan, Cortidelt, Dacorten, Dacortin, Decortancyl, Decortin, Decortisyl, Dehydrocortisone, Dekortin, Delcortin, Dellacort, Dellacort A, Delta E, Delta-Cortelan, Delta-Dome, Deltacortene, Deltacortisone, Deltacortone, Deltasone, Deltison, Deltisona, Deltisone, Deltra, Di-Adreson, Drazone, Econosone, Encorton, Encortone, Enkorton, Fernisone, Hostacortin, Liquid Pred, Me-Korti, Metacortandracin, Meticorten, NSC 10023, Nisona, Nurison, Orasone, Panafcort, Panasol, Paracort, Parmenison, Pehacort, Prednicen M, Prednicorm, Prednicort, Prednicot, Prednidib, Prednilonga, Predniment, Prednison, Prednitone, Pronison, Pulmison, Rectodelt, Retrocortine, Sone, Sterapred, Supercortil, Ultracorten, Ultracortene, Uniplen, Winpred, 1,4-Pregnadiene-17alpha,21-diol-3,11,20-trione, 17alpha,21-Dihydroxy-1,4-pregnadiene-3,11,20-trione, Delta-Cortisone, Delta1-Cortisone, Delta1-Dehydrocortisone, Prednisolone Impurity B |
| InChI Formula | InChI=1S/C21H26O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-15,18,22,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,18+,19-,20-,21-/m0/s1 |
| SMILES | C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO |
| IUPAC Name | (8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,11-dione |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction