Learn More
Uvex Preformed Mask silv-Air c 2312 FFP3
Product Code. 17532965 Shop All Uvex Products
Change view
Click to view available options
:
Filter Type: FFP3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17532965 Filter Type: FFP3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17532965 Supplier Uvex Supplier No. 8732312
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
The silv-Air c product family consists of classic folding and cup-shaped face masks which fit different face shapes. Headband lacing system and integrated sealing lips in all protection classes make it easy to fit these masks correctly – and to wear them comfortably for long periods.
General Features
  • Particle-filtering preformed mask with exhalation valve – ergonomically adapted for smaller faces
  • Color: white
Protection Features
  • Certified FFP3 face mask to EN 149:2001+A1:2009
  • D label (dolomite dust test passed): maximum breathability even with really high dust levels
  • Cannot be reused
Comfort Features
  • Four-point headband attachment and flexible, adjustable nose clip for a comfortable, secure fit
  • Very soft, all-around sealing lip for optimal wearer comfort and secure fit
  • Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
  • Ergonomically adjusted especially for smaller to medium faces
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 149 (D marked)
Design Type Cup/Molded
Disposable Single-use
Material Polypropylene, Polyester
Size Small/Medium
With Exhalation Valve Yes
Filter Efficiency FFP3
Sterility Non-sterile