Learn More
Uvex Preformed Mask silv-Air p 7220 FFP2
Product Code. 17502955 Shop All Uvex Products
Change view
Click to view available options
:
Filter Type: FFP2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17502955 Filter Type: FFP2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17502955 Supplier Uvex Supplier No. 8707220
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
The gray coloring on the outside of the silv-Air p masks prevents them from looking dirty quickly and having to be disposed of prematurely by the user. By contrast, the inside is of course pure white. In addition and with all products in this family, the 3D nose geometry ensures an optimum fit, even with different face shapes.
General Features
  • Particle-filtering preformed mask with exhalation valve to reduce exhalation resistance and heat and moisture build-up inside the mask
  • Metal-free processing
  • Additional activated carbon filter against odors, gases and vapors below threshold values
  • Seamless headband for a comfortable, secure fit
  • Color: silver, anthracite
Protection Features
  • FFP2 respirator certified in accordance with EN 149:2001+A1:2009
  • D marking (mask has passed the dolomite dust test): good breathability even with high levels of dust
  • Not reusable
Comfort Features
  • Seamless headband for a comfortable, secure fit
  • Exhalation valve to reduce breathing resistance and minimise heat and moisture build-up inside the mask
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 149 (D marked)
Disposable Single-use
Material Polypropylene, Polyester
Product Type Preformed with Carbon Filter
Size Standard
With Exhalation Valve Yes
Filter Efficiency FFP2
Sterility Non-sterile