missing translation for 'onlineSavingsMsg'
Learn More

Pregabalin, Lipomed

Product Code. 30457536
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457536 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457536 Supplier Lipomed Supplier No. LPMPRE147550

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Pregabalin
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pregabalin
CAS 148553-50-8
Analyte or Component Names Pregabalin
Shipping Condition Room Temperature
Molecular Formula C8 H17 N O2
Synonym Hexanoic acid, 3-(aminomethyl)-5-methyl-, (3 S)-, Hexanoic acid, 3-(aminomethyl)-5-methyl-, (S)-, (2 S)-3-(Aminomethyl)-5-methylhexanoic acid, (3 S)-3-(Aminomethyl)-5-methylhexanoic acid, 3 S-(aminomethyl)-5-methyl-hexanoic acid, (S)-(+)-3-(Aminomethyl)-5-methylhexanoic acid, (S)-3-(Ammoniomethyl)-5-methylhexanoate, (S)-Pregabalin, CI 1008, Gabanext 75, Lyrica, Mahagaba-75, Maxgalin 75, Neugaba 75, PD 144723, Pregabalin, Pregalin 75, (S)-Isobutylgaba
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CC(C)C[C@H](CN)CC(=O)O
IUPAC Name (3 S)-3-(aminomethyl)-5-methylhexanoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 159.23
Quantity 50 mg
Formula Weight 159.1259 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.