Learn More
PRELID2 Mouse anti-Human, Clone: OTI2F6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF810771
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
PRELID2 (PRELI Domain Containing 2) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include phosphatidic acid transporter activity. An important paralog of this gene is PRELID3B.Specifications
| PRELID2 | |
| Monoclonal | |
| Unconjugated | |
| PRELID2 | |
| PRELI domain containing 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 153768 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2F6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8N945 | |
| PRELID2 | |
| Full length human recombit protein of human PRELID2 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.