Description
The PM full Kit is designed to keep you running with high performance while ensuring long life and reliability, even under high workloads.
- Reduces unplanned downtime with consequent improvement of profit and production performance
- Lowers costs for the purchase of necessary spare parts and eventual extraordinary maintenance
- Reduces lost time associated with unplanned service intervals and instrument downtime
- Includes all necessary spare parts from the manufacturer
- Preventative maintenance needs to be carried out by a VELP™ authorized technician to guarantee durability, and performance per VELP™ quality standards
Specifications
Tekniske data
| For Use With (Equipment) | CSF6 Filtration Unit |
| Product Type | Maintenance Kit |
| Includes | 18 x EPDM CSF6 COEX Gaskets, 12 x OR 4131 FKM, 10 x T-Piece Union (6 mm Dia.), 5 x L-Piece Union (6 mm Dia.), 6 x Disk Wire Mesh, Norprene Tube (7.9 x 11.1 mm Dia.), Set of CSF6 Tubes |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?