missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Primidone-d5 (ethyl-d5), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Primidone-d5 |
| CAS | 73738-06-4 |
| Molecular Formula | C12 2H5 H9 N2 O2 |
| Quantity | 0.005 g |
| InChI Formula | InChI=1 S/C12H14N2O2/c1-2-12(9-6-4-3-5-7-9)10(15)13-8-14-11(12)16/h3-7 H,2,8H2,1H3,(H,13,15)(H,14,16)/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C1(C(=O)NCNC1=O)c2ccccc2 |
| IUPAC Name | 5-(1,1,2,2,2-pentadeuterioethyl)-5-phenyl-1,3-diazinane-4,6-dione |
| Molecular Weight (g/mol) | 223.28 |
| Formula Weight | 223.1369 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction