Description
- External thermal printer for cycle documentation.
- During the cycle the printer records temperature, pressure, date, cycle status, end of cycle.
Specifications
Specifications
| For Use With (Equipment) | D Line Autoclaves |
| Product Type | Printer |