missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olympus™ Prism, U-DICTS
Description
- DIC prism.
- Improves clarity and contrast for the following:
- General specimens.
- Tissue slices.
- Color-coded: White
Specifications
Specifications
| Color | White |
| Description | For visualizing general specimens, tissue slices. |
| For Use With (Application) | Visualizing general specimens, tissue slices. |
| For Use With (Equipment) | Biological microscopes. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?