missing translation for 'onlineSavingsMsg'
Learn More

Pritelivir, Thermo Scientific™

Product Code. 17535585 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17535585 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 17535585 Supplier Thermo Scientific Acros Supplier No. 469231000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Specifications

CAS 348086-71-5
Quantity 100 mg
Molecular Formula C18 H18 N4 O3 S2
Formula Weight 402.5
MDL Number MFCD18633192
Physical Form Powder
Color White
Purity ≥94%
InChI Key IVZKZONQVYTCKC-UHFFFAOYSA-N
SMILES CN(C(=O)CC1=CC=C(C=C1)C1=CC=CC=N1)C1=NC(C)=C(S1)S(N)(=O)=O
IUPAC Name N-methyl-N-(4-methyl-5-sulfamoyl-1,3-thiazol-2-yl)-2-[4-(pyridin-2-yl)phenyl]acetamide
Molecular Weight (g/mol) 402.49
Chemical Name or Material Pritelivir
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.